Human IL36 beta (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Interleukin-36 beta
- Protein/peptide name:IL36 beta
- Purity:> 97 % by SDS-PAGE and HPLC analyses
- Molecular weight:17.7kDa
- Sequence:MNPQREAAPK SYAIRDSRQM VWVLSGNSLI AAPLSRSIKP VTLHLIACRD TEFSDKEKGNMVYLGIKGKD LCLFCAEIQG KPTLQLKEKN IMDLYVEKKA QKPFLFFHNK EGSTSVFQSVSYPGWFIATS TTSGQPIFLT KERGITNNTN FYLDSVE
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together