Rat recombinant GDNF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Glial Cell-derived Neurotrophic Factor
- Protein/peptide name:GDNF
- Purity:> 95 % by SDS-PAGE and HPLC analyses
- Molecular weight:15.2 kDa
- Sequence:MSPDKQAAAL PRRERNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLNVTDLGLGYET KEELIFRYCS GSCEAAETMY DKILKNLSRS RRLTSDKVGQACCRPVAFDD DLSFLDDSLV YHILRKHSAK RCGCI
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, 500 mM NaCl, pH 6.4
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together