HCG Ag
BIRBORB98822-0.5
: Biorbyt
:
- Pk:0,5 mg
- Storage conditions:Store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/peptide name:HCG Ag
- Purity:Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
- Formulation:PBS, pH 7.4
- Tested applications:ELISA