Mouse TISMP (from skeletal muscle)
BIRBORB79713-0.1
: Biorbyt
:
- Pk:0,1 mg
- Source:Skeletal muscle
- Species:Mouse
- Storage conditions:Store at - 20 °C for short term and at - 70 °C for long term storage
- Protein/peptide name:TISMP
- Purity:95 % (SDS-PAGE) purified
- Solubility:Solubility is limited at neutral pH and physiological salt concentration
- Formulation:Freeze-dried from 0.01 M HCl