Recombinant IL1RA
Catalog # BIRBORB97791-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: 0.5 ng/ml, corresponding to a specific activity of 2x106 IU/mg determined by the dose dependent stimulation of murine D10S cells.
- Protein synonyms:DIRA
- UniProtKB:P18510
- Protein/peptide name:IL1RA
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized IL-1 ra insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions. Forlong term storage it is recommended to add a carrierprotein (0.1% HSA or BSA).
- Molecular weight:17 kDa
- Formulation:Lyophilized with no additives.