Order Entry
Portugal
ContactUsLinkComponent
Borrelia burgdorferi Recombinant Osp-A (from E. coli)
Borrelia burgdorferi Recombinant Osp-A (from E. coli)
Catalog # BIRBORB82568-100
Supplier:  Biorbyt
CAS Number:  
Borrelia burgdorferi Recombinant Osp-A (from E. coli)
Catalog # BIRBORB82568-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Borrelia burgdorferi
  • Protein synonyms:
    B burgdorferi
  • Protein/peptide name:
    Osp-A
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    WB

Specifications

About this item