Human p53 Mutant (from E. coli)
Marketsource
: Biorbyt
- Catalog No:
- BIRBORB81252-2
- BIRBORB81252-5
- BIRBORB81252-10
- Protein/peptide name:
- p53 Mutant
- p53 Mutant
- p53 Mutant
- Protein synonyms:
- BCC7
- BCC7
- BCC7
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Formulation:
- p53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- p53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- p53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Storage conditions:
- Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Tested applications:
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
Product Family Options
- Pk
- Marketsource
Specifications
Cat. No.BIRBORB81252-2Protein/peptide namep53 MutantProtein synonymsBCC7SpeciesHumanSourceE. coliFormulationp53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.Storage conditionsStore vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.Tested applicationsSDS-PAGEMarketsource - Marketsource
Specifications
Cat. No.BIRBORB81252-5Protein/peptide namep53 MutantProtein synonymsBCC7SpeciesHumanSourceE. coliFormulationp53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.Storage conditionsStore vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.Tested applicationsSDS-PAGEMarketsource - Marketsource
Specifications
Cat. No.BIRBORB81252-10Protein/peptide namep53 MutantProtein synonymsBCC7SpeciesHumanSourceE. coliFormulationp53 Mutant at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.Storage conditionsStore vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.Tested applicationsSDS-PAGEMarketsource



