Order Entry
Portugal
ContactUsLinkComponent
Anti-CYCS Mouse Monoclonal Antibody [clone: CYCS/1010]
Anti-CYCS Mouse Monoclonal Antibody [clone: CYCS/1010]
Catalog # ABNOMAB14395
Supplier:  Abnova
Anti-CYCS Mouse Monoclonal Antibody [clone: CYCS/1010]
Catalog # ABNOMAB14395
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Recombinant CYCS
  • Antigen symbol:
    CYCS
  • Clonality:
    Monoclonal
  • Clone:
    CYCS/1010
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    54205
  • Amino acid number:
    P99999
  • Storage buffer:
    In 1 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Immunogen:
    Recombinant protein corresponding to full length human CYCS.
  • Purification:
    Protein A/G purification
  • Size:
    100 µG
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant human CYCS.

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: CYCS/1010
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human