Anti-APRIL Rabbit Polyclonal Antibody
Catalog # ENZOADICSA836E
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:A Proliferation-inducing Ligand
- Antigen symbol:APRIL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:APRIL|TNFSF13|TALL2|TRDL-1|CD256|UNQ383/PRO715
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:4 °C
- Shipping temperature:Blue ice not frozen
- Immunogen:Synthetic peptide corresponding to a portion of human APRIL.
- Purification:Purified by peptide affinity chromatography
- Pk:100 µG
Specifications
About this item
APRIL (A proliferation-inducing ligand) is a member of the TNF family. APRIL stimulates B- and T-cell proliferation, triggers humoral immune responses, activates NF-κB, and induces cell death.
- Dilution range: Immunohistochemistry (10 µg/ml), Western blot (1-4 µg/ml)
- Detects a band of ~27kDa by Western blot
Type: Primary
Antigen: APRIL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse