Potassium bromide ≥99.0%, white crystalline powder ACS
:7758-02-3
:
:KBr
:MFCD00011358
:~734 °C (lit.)
:119 g/mol
:GHS07
- Identity Test:Passes
- Assay:≥99,0%
- pH (5% aq soln):5,0 - 8,8
- Insoluble Matter:<0,005%
- Bromate (BrO3):<0,001%,
- Iodate (IO3):<0,001%
- Chloride (cl):<0,2%
- Iodide (I):<0,001%
- Sulfate (SO4):<0,005%
- Barium (Ba):<0,002%
- Heavy Metals (as Pb):<5 ppm
- Iron (Fe):<5 ppm
- Calcium (Ca):<0,002%
- Magnesium (Mg):<0,001%,
- Sodium (Na):<0,02%
Frequently Bought Together