Calcium sulphate dihydrate ≥98.0%, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:10101-41-4
:
Formula:CaSO₄·2H₂O
MDL Number:MFCD00149625
:172.17 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay:≥98,0%
- Insoluble in Dilute Hydrochloric Acid:≤0,02%
- Chloride:≤0,005%
- Nitrate:Passes
- Carbonate:Passes
- Heavy Metals:≤0,002%
- Iron:<0,001%
- Magnesium:≤0,02%
- Potassium:≤0,005%
- Sodium:≤0,02%
- Strontium:≤0,05%
Specification Test Results
Frequently Bought Together