Order Entry
Portugal
ContactUsLinkComponent
Bovine carbonic anhydrase (from Erythrocytes), MP Biomedicals
Bovine carbonic anhydrase (from Erythrocytes), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Bovine carbonic anhydrase (from Erythrocytes), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Source:
    Erythrocytes
  • Species:
    Bovine
  • Storage conditions:
    2...8 °C
  • Form:
    Tan powder
  • Enzyme name:
    Carbonic anhydrase
  • Enzyme activity:
    >2000 u/mg solid
  • Enzyme synonyms:
    E.C. 4.2.1.1, Carbonate Dehydratase, Carbonate Hydrolyase

Specifications

Frequently Bought Together