Order Entry
Portugal
ContactUsLinkComponent
Ammonium iodide, powder ACS
Ammonium iodide, powder ACS
Supplier:  MP Biomedicals
Certificates
Ammonium iodide, powder ACS
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Form:
    Powder
  • Storage:
    Store at Room Temperature (15 - 30 °C), Desiccate, Protect from light
  • Assay:
    ≥99.0%
  • Heavy Metals:
    ≤0,001%
  • Insoluble Matter:
    ≤0,005%
  • Residue on Ignition:
    ≤0,05%
  • Chloride and Bromide:
    ≤0,005%
  • Phosphate:
    ≤0,001%
  • Sulfate:
    ≤0,05%
  • Barium:
    ≤0,002%
  • Iron:
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together