Anti-PTPRC Mouse Monoclonal Antibody (PerCP) [clone: D3/9]
Catalog # ABNOMAB13931
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:D3/9
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:5788
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human PTPRC.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: D3/9
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human