Anti-CD34 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 581]
ABNOMAB13882
: Abnova
- Antibody type:Primary
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:581
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:947
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against human CD34.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human