Anti-ITGA2B Mouse Monoclonal Antibody (FITC) [clone: HIP8]
Catalog # ABNOMAB13899
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:HIP8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:3674
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human ITGA2B.
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: HIP8
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human