Anti-TGFA6 Mouse Monoclonal Antibody [clone: 1E8-G6]
Catalog # ABNOMAB13444
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:TGFA6
- Clonality:Monoclonal
- Clone:1E8-G6
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Gene ID:7039
- Sequence:PPVAAAVVSH
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of human TGFA.
Type: Primary
Antigen: TGFA6
Clonality: Monoclonal
Clone: 1E8-G6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human