Calcium bromide hydrate, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71626-99-8
:
Formula:Br₂Ca·H₂O
MDL Number:MFCD00149608
:217.9 g/mol
Melting Pt:730 °C
Density:2,295 g/cm³ (20 °C)
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder and/or chunks
- Titration Complexometric:≥82 % (CaBr2)
- Solubility:(10 % in water) Clear colorless to slightly hazy
- Acidity/alkalinity:Passes test
Specification Test Results
Frequently Bought Together