Calcium carbonate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Melting Pt:825 °C (dec.)
Density:2,71 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:39 to 42 %
- Total trace metal impurities:≤10 ppm
Specification Test Results
Frequently Bought Together