Isoindoline hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32372-82-0
:
Formula:C₈H₁₀ClN
MDL Number:MFCD03502377
:155.625 g/mol
Melting Pt:255…256 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white to grey Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Elemental Analysis:59.27 to 64.21 % C
Specification Test Results
Frequently Bought Together