Order Entry
Portugal
ContactUsLinkComponent
 
 
2-Cyano-6-methylpyridine 97%
 
undefined
 
2-Cyano-6-methylpyridine 97%

 

  • Appearance:
    White to orange to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 73 °C
  • GC:
    ≥96 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

Frequently Bought Together