
DL-Alanine methyl ester hydrochloride 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13515-97-4
:
Formula:C₄H₉NO₂
MDL Number:MFCD00035523
:139.58 g/mol
Melting Pt:157…158 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥98.0 %
Specification Test Results
Frequently Bought Together