(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:167834-24-4
:
Formula:C₁₁H₁₆ClNO₂
MDL Number:MFCD02259715
:229.704 g/mol
Melting Pt:117 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
Frequently Bought Together