Poly(acrylic acid), M.W. ∼ 240,000 25% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-01-4
:9062-04-8
Formula:C₃H₄O₂
MDL Number:MFCD00084394
:72.063 g/mol
Boiling Pt:100 °C (760 mmHg)
Density:1.09 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Assay:23 to 27 % Solids
- Molecular weight:240000 (approx)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together