
3,4-Dichlorobenzyl isocyanate 98%
:19752-09-1
:
:C₈H₅Cl₂NO
:MFCD00673063
:202.034 g/mol
:282 °C (760 mmHg)
:1.375 g/cm³
:>110 °C
:Ambient
:2206
:6.1,III
:GHS07
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5656 to 1.5676 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together