4-Aminobutyric acid
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:56-12-2
:
Formula:C₄H₉NO₂
MDL Number:MFCD00008226
:103.12 g/mol
Merck Index:13,00429
Boiling Pt:248 °C (1013 hPa)
Melting Pt:202 °C
Density:1.21 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-258-6
Specification Test Results
- Assay:98% Min.
Specification Test Results
Frequently Bought Together