
2-(Hydroxymethyl)pyridine-5-boronic acid 97%
Catalog # H53319.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:152.94 g/mol
:GHS07
CAS Number:913835-98-0
Formula:C₆H₈BNO₃
MDL Number:MFCD09027230