Rhodium 5%, reduced Type 526
Catalog # 47120.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:102.9 g/mol
Storage Temperature:Ambient
EINECS:231-125-0
CAS Number:7440-16-6
Formula:Rh
MDL Number:MFCD00011201