Order Entry
Portugal
ContactUsLinkComponent
Accessories for Medifuge™ centrifuge
undefined
Accessories for Medifuge™ centrifuge
  • Catalog No:
  • 521-2228
  • 521-2231
  • 521-2230
  • 521-2232
  • 521-2229
  • 521-2233
  • Max. speed:
  • 300 - 4900 rpm
  • Max. RCF:
  • Max. RCF - fixed angle rotor:
  • 3114 ×g
  • Max. RCF - swing-out rotor:
  • 3490 ×g
  • Speed increment:
  • 100 rpm
  • Capacity:
  • 8×15 ml (fixed angle), 8×10 ml (swinging bucket)
Frequently Bought Together