Anti-IGF1R Rabbit Polyclonal Antibody
Catalog # ABNOPAB29290
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Insulin-Like Growth Factor 1 Receptor
- Antigen symbol:IGF1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3480
- Antigen synonyms:JTK13|CD221|IGFR|IGFIR
- Modification:phospho-Tyr1346
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y1346 of human IGF1R.
- Purification:purified by Affinity Chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human IGF1R.
Recommended Dilutions: Immunohistochemistry: 1:50~1:100
Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: