Order Entry
Portugal
ContactUsLinkComponent
Anti-HRASLS5 Mouse Monoclonal Antibody [clone: 2F5]
Anti-HRASLS5 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00117245-M02
Supplier:  Abnova
Anti-HRASLS5 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00117245-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HRAS-like suppressor family, member 5
  • Antigen symbol:
    HRASLS5
  • Clonality:
    Monoclonal
  • Clone:
    2F5
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    117245
  • Antigen synonyms:
    HRSL5|iNAT|RLP1|HRLP5
  • Amino acid number:
    1 to 269
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MGLSPGAEGEYALRLPRIPPPLPKPASRTAGTGPKDQPPALRRSAVPHSEESVGFAALVQLPAKQPPPGTLEQGRSIQQGEKAVVSLETTPSQKADWSSIPKPENEGKLIKQAAEGKPRPRPGDLIEIFRIGYEHWAIYVEDDCVVHLAPPSEEFEVGSITSIFSNRAVVKYSRLEDVLHGCSWKVNNKLDGTYLPLPVDKIIQRTKKMVNKIVQYSLIEGNCEHFVNGLRYGVPRSQQVEHALMEGAKAAGAVISAVVDSIKPKPITA
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    HRASLS5 (AAH34222.1, 1 a.a. ~ 269 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a full-length recombinant HRASLS5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HRASLS5
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: