Order Entry
Portugal
ContactUsLinkComponent
 
Anti-Myeloid Cell Marker Mouse Monoclonal Antibody [clone: SPM298]
  ABNOMAB12081
 :  Abnova
Anti-Myeloid Cell Marker Mouse Monoclonal Antibody [clone: SPM298]
  ABNOMAB12081
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Myeloid Cell Marker
  • Antigen symbol:
    Myeloid Cell Marker
  • Clonality:
    Monoclonal
  • Clone:
    SPM298
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,05% BSA, 0,05% sodium azide).
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Human peripheral blood mononuclear cells.
  • Purification:
    Protein A/G purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against Myeloid Cell Marker.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: Myeloid Cell Marker
Clonality: Monoclonal
Clone: SPM298
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: