Anti-COL19A1 Mouse Monoclonal Antibody [clone: 1B2]
Catalog # ABNOH00001310-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Collagen, type XIX, alpha 1
- Antigen symbol:COL19A1
- Clonality:Monoclonal
- Clone:1B2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1310
- Antigen synonyms:COL9A1L|D6S228E
- Amino acid number:27 to 114
- Storage buffer:1x PBS, pH 7,4
- Sequence:RDKTEESCPILRIEGHQLTYDNINKLEVSGFDLGDSFSLRRAFCESDKTCFKLGSALLIRDTIKIFPKGLPEEYSVAAMFRVRRNAKK
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:COL19A1 (NP_001849, 27 a.a. ~ 114 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant COL19A1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: COL19A1
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: