Anti-CK18 Mouse Monoclonal Antibody (Biotin) [clone: DA-7]
ABNOMAB6506
: Abnova
- Antibody type:Primary
- Antigen name:Cytokeratin 18
- Antigen symbol:CK18
- Clonality:Monoclonal
- Clone:DA-7
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:3875
- Antigen synonyms:K18|CYK18|CK-18
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified KRT18 from human breast carcinoma cell line (PMC-42).
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native KRT18.
Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: DA-7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human