Order Entry
Portugal
ContactUsLinkComponent
Culture media, in tubes
Culture media, in tubes
Catalog # HEIP1463030100
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
undefined
Culture media, in tubes
Catalog # HEIP1463030100
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Specifications
  • Pk:
    100
  • Standards:
    EP, USP
  • Description:
    Sabouraud dextrose agar
  • Filling:
    18 ml
Specifications
About this item
Sabouraud dextrose is recommended for the microbial enumeration tests.