Order Entry
Portugal
ContactUsLinkComponent
 
Anti-Complement C3a, C3a des-Arg, Human Mouse Monoclonal Antibody [clone: 6F68]
  USBIC7850-13H
Anti-Complement C3a, C3a des-Arg, Human Mouse Monoclonal Antibody [clone: 6F68]
  USBIC7850-13H

 

  • Antibody type:
    Primary
  • Antigen name:
    Complement C3a, C3a des-Arg, Human
  • Clonality:
    Monoclonal
  • Clone:
    6F68
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide before the addition of glycerol to 40%.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: Complement C3a, C3a des-Arg, Human
Clonality: Monoclonal
Clone: 6F68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human