Anti-BrdU (Ab-3) (Mobu-1) Mouse Monoclonal Antibody
NA61-100
- Antibody type:Primary
- Antigen symbol:BRDU
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: BrdU (Ab-3) (Mobu-1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan