Order Entry
Portugal
ContactUsLinkComponent
 
Anti-Oval Cell Marker Rat Monoclonal Antibody (Biotin) [clone: OC2-6E10]
  NOVUNBP1-18981B
Anti-Oval Cell Marker Rat Monoclonal Antibody (Biotin) [clone: OC2-6E10]
  NOVUNBP1-18981B

 

  • Antibody type:
    Primary
  • Antigen name:
    Oval Cell Marker
  • Clonality:
    Monoclonal
  • Clone:
    OC2-6E10
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-6E10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse