Specifications
- Antibody type:Primary
- Antigen symbol:HCG Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ULG5|LCGR|LH/CG-R|LHR|HHG|LSH-R|LHRHR|LH/CGR|LGR2
- Amino acid number:127 to 143
- UniProtKB:P22888
- Sequence:YLSICNTGIRKFPDVTK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human hCG receptor (127-143aa YLSICNTGIRKFPDVTK), different from the relative rat sequence by two amino acids, mouse sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Lutropin-choriogonadotropic hormone receptor(LHCGR) detection. Tested with WB, IHC-P in Human;Rat.
The luteinizing hormone/choriogonadotropin receptor is a member of a subfamily of G protein-coupled receptors(GPCR) characterized by the presence of a large N-terminal extracellular domain containing several leucine-rich repeats(LRR). This glycoprotein hormone receptor family has been named the LRR-containing GPCR(LGR) family. The LHCGR gene contains 11 exons and spans approximately 80 kb and it is mapped to chromosome 2p21. In the ovary, theca, stromal, late-stage(luteinizing) granulosa, and luteal cells contain LHCGR. In the testes, Leydig cells contain LHCRG.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: hCG Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat