Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Supplier: Biotium
Certificates
About this item
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Specifications
- Catalog No:
- 20104-1
- 20104.
- Antigen symbol:
- IgG
- IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 568
- CF® 568
- Reactivity:
- Chicken
- Chicken
- Host:
- Goat
- Goat
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together