Order Entry
Portugal
ContactUsLinkComponent
Anti-Desmin K5 Rabbit Polyclonal Antibody
Anti-Desmin K5 Rabbit Polyclonal Antibody
Catalog # BIRBORB98300-0.25
Supplier:  Biorbyt
Anti-Desmin K5 Rabbit Polyclonal Antibody
Catalog # BIRBORB98300-0.25
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Desmin K5
  • Antigen symbol:
    DES
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage buffer:
    PBS containing 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    250 μL
  • Pk:
    0,25 mL

Specifications

About this item

Type: Primary
Antigen: Desmin K5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: