Anti-EPM2A Rabbit Polyclonal Antibody
Catalog # BIRBORB96249-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Epilepsy progressive myoclonus type 2A Lafora disease (laforin)
- Antigen symbol:EPM2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 0.25-0.5 ?g/ml
Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: