Anti-XPR1 Rabbit Polyclonal Antibody
Catalog # BIRBORB85035-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:xenotropic and polytropic retrovirus receptor 1
- Antigen symbol:XPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 16 amino acid peptide from N-terminal extracellular domain of human XPR1
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunofluorescence: 8-15 ?g/ml; Immunochemistry: 8 ?g/ml
Type: Primary
Antigen: XPR1 (xenotropic and polytropic retrovirus receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human