Order Entry
Portugal
ContactUsLinkComponent
Anti-XPR1 Rabbit Polyclonal Antibody
Anti-XPR1 Rabbit Polyclonal Antibody
Catalog # BIRBORB79864-50
Supplier:  Biorbyt
undefined
Anti-XPR1 Rabbit Polyclonal Antibody
Catalog # BIRBORB79864-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    xenotropic and polytropic retrovirus receptor 1
  • Antigen symbol:
    XPR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic 16 amino acid peptide from C-terminus of human XPR1
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Immunochemistry: 8 ?g/ml

Type: Primary
Antigen: XPR1 (xenotropic and polytropic retrovirus receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human