Order Entry
Portugal
ContactUsLinkComponent
Anti-BAX Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anti-BAX Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB124471-100
Supplier:  Biorbyt
Anti-BAX Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB124471-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCL2-associated X Protein
  • Antigen symbol:
    BAX
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    21 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Bax C-terminus.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: