Anti-LCP2 Mouse Monoclonal Antibody
Catalog # BIRBORB44776-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:lymphocyte cytosolic Protein 2 (SH2 domain containing leukocyte Protein of 76kDa)
- Antigen symbol:LCP2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: