Order Entry
Portugal
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB8147-100
Supplier:  Biorbyt
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB8147-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD19 molecule
  • Antigen symbol:
    CD19
  • Clonality:
    Monoclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    PBS, 50% glycerol, 0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human