Anti-SAP30 Mouse Polyclonal Antibody
ABNOH00008819-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Sin3A-associated Protein, 30kDa
- Antigen symbol:SAP30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8819
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: SAP30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human