Anti-BANF1 Mouse Monoclonal Antibody [clone: M2]
Catalog # ABNOH00008815-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Barrier To Autointegration Factor 1
- Antigen symbol:BANF1
- Clonality:Monoclonal
- Clone:M2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8815
- Antigen synonyms:NGPS|BCRG1|Barrier-to-autointegration factor|BCRP1|BANF1|Breakpoint cluster region protein 1|D14S1460|BAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BANF1
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human