Anti-UCHL3 Mouse Monoclonal Antibody [clone: 4E9]
Catalog # ABNOH00007347-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal esterase L3
- Antigen symbol:UCHL3
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7347
- Antigen synonyms:Ubiquitin thioesterase L3|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN.|Ubiquitin thiolesterase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UCHL3
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat